Skip to Content
Merck
HEPES solution BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered

HEPES solution

HEPES solution BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Synonym(s):
N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), HEPES solution
CAS Number:

Products

Product Category
Brand
Color
Feature
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Sterilization Method
Technique