![Halothane United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/260/701/a7ca9d23-b745-4ece-9474-d3a1547bfdaa/640/a7ca9d23-b745-4ece-9474-d3a1547bfdaa.png)
Halothane
![Halothane United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/260/701/a7ca9d23-b745-4ece-9474-d3a1547bfdaa/640/a7ca9d23-b745-4ece-9474-d3a1547bfdaa.png)
Synonym(s):
2-Bromo-2-chloro-1,1,1-trifluoroethane, Halothane
Linear Formula:
BrCH(Cl)CF3
CAS Number:
Molecular Weight:
197.38
Products
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing