Glyburide Related Compound B
Synonym(s):
Methyl {{4-{2-[(5-chloro-2-methoxybenzoyl)amino]ethyl}phenyl}sulfonyl}carbamate
Empirical Formula (Hill Notation):
C18H19ClN2O6S
CAS Number:
Molecular Weight:
426.87
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2