![Glucoiberin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/233/255/26515c9e-83f6-409a-ba8f-11dcbd5d769c/640/26515c9e-83f6-409a-ba8f-11dcbd5d769c.png)
Glucoiberin potassium salt
![Glucoiberin potassium salt phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/233/255/26515c9e-83f6-409a-ba8f-11dcbd5d769c/640/26515c9e-83f6-409a-ba8f-11dcbd5d769c.png)
Synonym(s):
3-(Methylsulfinyl)propylglucosinolate potassium salt
Empirical Formula (Hill Notation):
C11H20KNO10S3
CAS Number:
Molecular Weight:
461.57
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing