Skip to Content
Merck
Gentamicin sulfate European Pharmacopoeia (EP) Reference Standard

Gentamicin sulfate

Gentamicin sulfate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Gentamicin sulfate salt
CAS Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Manufacturer
Physical Form
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique