Skip to Content
Merck
γ-Decalactone analytical standard

γ-Decalactone

γ-Decalactone analytical standard
Synonym(s):
γ-Decanolactone, 4-Hydroxydecanoic acid γ-lactone
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing