![Fluconazole Impurity A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/599/257/3d0278f5-62a1-49e3-b8aa-19fd72809ce8/640/3d0278f5-62a1-49e3-b8aa-19fd72809ce8.jpg)
Fluconazole Impurity A
![Fluconazole Impurity A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/599/257/3d0278f5-62a1-49e3-b8aa-19fd72809ce8/640/3d0278f5-62a1-49e3-b8aa-19fd72809ce8.jpg)
Synonym(s):
(2RS)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)-3-(4H-1,2,4-triazol-4-yl)propan-2-ol
Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing