Skip to Content
Merck
Floxuridine pharmaceutical secondary standard, certified reference material

Floxuridine

Floxuridine pharmaceutical secondary standard, certified reference material
Synonym(s):
5-Fluoro-2′-deoxyuridine, 2′-Deoxy-5-fluorouridine, FUDR, Floxuridine
Empirical Formula (Hill Notation):
C9H11FN2O5
CAS Number:
Molecular Weight:
246.19

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade