![Eplerenone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/563/759/22f7f86f-5ddd-4700-b9b3-10a9cbd05285/640/22f7f86f-5ddd-4700-b9b3-10a9cbd05285.png)
Eplerenone for peak identification
![Eplerenone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/563/759/22f7f86f-5ddd-4700-b9b3-10a9cbd05285/640/22f7f86f-5ddd-4700-b9b3-10a9cbd05285.png)
Synonym(s):
Eplerenone, 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic acid, gamma-lactone, methyl ester, Cgp 30083, Eplerenone [USAN], Epoxymexrenone, HSDB 7522, Inspra, Pregn-4-ene-7,21-dicarboxylic acid, 9,11-epoxy-17-hydroxy-3-oxo-, gamma-lactone, methyl ester, (7alpha,11alpha,17alpha)-, SC-66110
Empirical Formula (Hill Notation):
C24H30O6
CAS Number:
Molecular Weight:
414.49
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing