Skip to Content
Merck
vibrant-m

Entecavir for system suitability

vibrant-m
Synonym(s):
2-Amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-6H-purin-6-one monohydrate
Empirical Formula (Hill Notation):
C12H15N5O3 · H2O
CAS Number:
Molecular Weight:
295.29

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1