![Endfitting w/ Fixed 10 μm Pyrex® Frit for 5 mm ID Glass TSKgel® Columns Pyrex frit, pkg of 1 ea](/deepweb/assets/sigmaaldrich/product/images/354/446/448a3121-c6f4-4b48-9cd3-68680825d2a1/640/448a3121-c6f4-4b48-9cd3-68680825d2a1.jpg)
Endfitting w/ Fixed 10 μm Pyrex® Frit for 5 mm ID Glass TSKgel® Columns
![Endfitting w/ Fixed 10 μm Pyrex® Frit for 5 mm ID Glass TSKgel® Columns Pyrex frit, pkg of 1 ea](/deepweb/assets/sigmaaldrich/product/images/354/446/448a3121-c6f4-4b48-9cd3-68680825d2a1/640/448a3121-c6f4-4b48-9cd3-68680825d2a1.jpg)
Synonym(s):
TSKgel® Hardware and Accessories
Products
Manufacturer
Product Line
Package Size
Pore size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing