Dulbecco’s Modified Eagle’s Medium/Ham’s Nutrient Mixture F12
Synonym(s):
Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham, DME/F-12, 1:1 mixture
Products
Product Category
Brand
Physical Form
L-Glutamine
Phenol Red
Quality Segment
Sterility
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing