Skip to Content
Merck
Doxazosin mesylate United States Pharmacopeia (USP) Reference Standard

Doxazosin mesylate

Doxazosin mesylate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[4-(1,4-benzodioxan-2-yl)carpiperazin-1-yl)]-6,7-dimethoxyquinazoline mesylate, Doxazosin methanesulfonate
Empirical Formula (Hill Notation):
C23H25N5O5 · CH3SO3H
CAS Number:
Molecular Weight:
547.58

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade