Skip to Content
Merck
Diphenyl sulfone PESTANAL®, analytical standard

Diphenyl sulfone

Diphenyl sulfone PESTANAL®, analytical standard
Synonym(s):
Phenyl sulfone
Linear Formula:
(C6H5)2SO2
CAS Number:
Molecular Weight:
218.27
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing