Skip to Content
Merck
Dimethyl disulfide analytical standard

Dimethyl disulfide

Dimethyl disulfide analytical standard
Synonym(s):
DMDS, Methyl disulfide, Dimethyl disulfide
Linear Formula:
CH3SSCH3
CAS Number:
Molecular Weight:
94.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6