
Diethyl azelate

Synonym(s):
Azelaic acid diethyl ester, Diethyl nonanedioate, Nonanedioic acid diethyl ester
Linear Formula:
C2H5O2C(CH2)7CO2C2H5
CAS Number:
Molecular Weight:
244.33
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing