Desoxycortone acetate
Synonym(s):
Deoxycorticosterone acetate, 11-Deoxycorticosterone acetate, 21-Acetoxy-4-pregnene-3,20-dione, 21-Acetoxyprogesterone, 21-Hydroxy-4-pregnene-3,20-dione 21-acetate, 21-Hydroxyprogesterone 21-acetate, 4-Pregnen-21-ol-3,20-dione 21-acetate, Cortexone acetate, DOC acetate
Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing