![D-2-Phosphoglyceric acid lithium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/412/162/99a6779e-fb7e-4f80-8aef-18ba9eda905c/640/99a6779e-fb7e-4f80-8aef-18ba9eda905c.png)
D-2-Phosphoglyceric acid lithium salt
![D-2-Phosphoglyceric acid lithium salt analytical standard](/deepweb/assets/sigmaaldrich/product/structures/412/162/99a6779e-fb7e-4f80-8aef-18ba9eda905c/640/99a6779e-fb7e-4f80-8aef-18ba9eda905c.png)
Synonym(s):
(2R)-3-Hydroxy-2-(phosphonooxy)propanoic acid lithium salt, Lithium D-2-phosphoglycerate
Empirical Formula (Hill Notation):
C3H7O7P · xLi+
CAS Number:
Molecular Weight:
186.06 (free acid basis)
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing