
Cyclophosphamide Related Compound D

Synonym(s):
3-{{2-[(2-Chloroethyl)amino]ethyl}amino}propyl monophosphate dihydrochloride
Empirical Formula (Hill Notation):
C7H18ClN2O4P · 2HCl
CAS Number:
Molecular Weight:
333.58
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing