![Clopyralid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/341/181/1dfbfd4f-100f-4157-8a5c-4fe64ca060bd/640/1dfbfd4f-100f-4157-8a5c-4fe64ca060bd.png)
Clopyralid
![Clopyralid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/341/181/1dfbfd4f-100f-4157-8a5c-4fe64ca060bd/640/1dfbfd4f-100f-4157-8a5c-4fe64ca060bd.png)
Synonym(s):
3,6-Dichloro-pyridine-2-carboxylic acid
Empirical Formula (Hill Notation):
C6H3Cl2NO2
CAS Number:
Molecular Weight:
192.00
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2