Skip to Content
Merck
Cloprostenol sodium United States Pharmacopeia (USP) Reference Standard

Cloprostenol sodium

Cloprostenol sodium United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(+/-)-Cloprostenol sodium salt hydrate, (+/-)-16-(3-Chlorophenoxy)-9,11,15R-trihydroxy-17,18,19,20-tetranorprosta-5Z,13E-dien-1-oic acid sodium salt hydrate
Empirical Formula (Hill Notation):
C22H28ClNaO6 · xH2O
CAS Number:
Molecular Weight:
446.90 (anhydrous basis)
EC Number:

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing