Skip to Content
Merck
vibrant-m

Citalopram Related Compound F

vibrant-m
Synonym(s):
(RS)-Dimethyl(1-methyl-3,3-diphenylallyl)amine hydrochloride
Empirical Formula (Hill Notation):
C18H21N · HCl
CAS Number:
Molecular Weight:
287.83

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1