![Cinosulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/237/957/a58176ae-0fa5-4062-9a67-a6860fc3a3d5/640/a58176ae-0fa5-4062-9a67-a6860fc3a3d5.png)
Cinosulfuron
![Cinosulfuron PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/237/957/a58176ae-0fa5-4062-9a67-a6860fc3a3d5/640/a58176ae-0fa5-4062-9a67-a6860fc3a3d5.png)
Empirical Formula (Hill Notation):
C15H19N5O7S
CAS Number:
Molecular Weight:
413.41
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing