Skip to Content
Merck
Cholest-5-en-26,26,26,27,27,27-d6-3-ol 98 atom % D, 97% (CP)

Cholest-5-en-26,26,26,27,27,27-d6-3-ol

Cholest-5-en-26,26,26,27,27,27-d6-3-ol 98 atom % D, 97% (CP)
Synonym(s):
Cholesterol-26,26,26,27,27,27-d6
Empirical Formula (Hill Notation):
C27D6H40O
CAS Number:
Molecular Weight:
392.69

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing