Cardamonin
Synonym(s):
(2E)-1-(2,4-Dihydroxy-6-methoxyphenyl)-3-phenyl-2-propen-1-one, (8CI); Alpinetin chalcone, (E)-2′,4′-Dihydroxy-6′-methoxy-chalcone
Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing