Carbidopa for system suitability
Synonym(s):
(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid monohydrate
Empirical Formula (Hill Notation):
C10H14N2O4 · H2O
CAS Number:
Molecular Weight:
244.24
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing