Skip to Content
Merck
Camphene primary reference standard

Camphene

Camphene primary reference standard
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing