Calcium chloride hexahydrate
Empirical Formula (Hill Notation):
CaCl2 · 6H2O
CAS Number:
Molecular Weight:
219.08
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing