Skip to Content
Merck
Caerulein ≥95% (HPLC)

Caerulein

Caerulein ≥95% (HPLC)
Synonym(s):
Caerulein Sulfated, Cerulein, Ceruletide, [Tyr(SO3H)4]Caerulein, pGlu-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2
Empirical Formula (Hill Notation):
C58H73N13O21S2
CAS Number:
Molecular Weight:
1352.40

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing