Skip to Content
Merck
Cadaverine analytical standard

Cadaverine

Cadaverine analytical standard
Synonym(s):
1,5-Diaminopentane, 1,5-Pentanediamine, Pentamethylenediamine
Linear Formula:
NH2(CH2)5NH2
CAS Number:
Molecular Weight:
102.18
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing