Buspirone for system suitability
Synonym(s):
Buspirone hydrochloride, N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride
Empirical Formula (Hill Notation):
C21H31N5O2 · HCl
CAS Number:
Molecular Weight:
421.96
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing