![Brij® C10 average Mn ~683](/deepweb/assets/sigmaaldrich/product/structures/575/955/fae093d0-105b-4b6f-9582-dbdb5c906098/640/fae093d0-105b-4b6f-9582-dbdb5c906098.png)
Brij® C10
![Brij® C10 average Mn ~683](/deepweb/assets/sigmaaldrich/product/structures/575/955/fae093d0-105b-4b6f-9582-dbdb5c906098/640/fae093d0-105b-4b6f-9582-dbdb5c906098.png)
Synonym(s):
Polyethylene glycol hexadecyl ether, Polyoxyethylene (10) cetyl ether
Linear Formula:
C16H33(OCH2CH2)nOH, n~10
CAS Number:
Products
Brand
Melting Point (°C)
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing