![Bis(1-butylpentyl) adipate Selectophore™, ≥98.0%](/deepweb/assets/sigmaaldrich/product/structures/144/831/f7f01c15-52c2-4744-be3a-dea9d3de248e/640/f7f01c15-52c2-4744-be3a-dea9d3de248e.png)
Bis(1-butylpentyl) adipate
![Bis(1-butylpentyl) adipate Selectophore™, ≥98.0%](/deepweb/assets/sigmaaldrich/product/structures/144/831/f7f01c15-52c2-4744-be3a-dea9d3de248e/640/f7f01c15-52c2-4744-be3a-dea9d3de248e.png)
Synonym(s):
Adipic acid di-5-nonyl ester, BBPA, Di(5-nonyl) adipate
Empirical Formula (Hill Notation):
C24H46O4
Products
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing