![beta-Sitosterol solution 200 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/422/879/af0013a3-bc33-441a-ad49-7d6c310ead83/640/af0013a3-bc33-441a-ad49-7d6c310ead83.png)
beta-Sitosterol solution
![beta-Sitosterol solution 200 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/422/879/af0013a3-bc33-441a-ad49-7d6c310ead83/640/af0013a3-bc33-441a-ad49-7d6c310ead83.png)
Synonym(s):
β-Sitosterol, α-Dihydrofucosterol, 22,23-Dihydrostigmasterol, 24α-Ethylcholesterol, 5-Stigmasten-3β-ol
Empirical Formula (Hill Notation):
C29H50O
CAS Number:
Molecular Weight:
414.71
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing