Skip to Content
Merck
Beta-Cyclodextrin Pharmaceutical Secondary Standard; Certified Reference Material

Beta Cyclodextrin

Beta-Cyclodextrin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
β-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin
Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique