Skip to Content
Merck
vibrant-m

Bendamustine Related Compound C

vibrant-m
Synonym(s):
Ethyl 4-(5-(bis(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoate
Empirical Formula (Hill Notation):
C18H27N3O4
CAS Number:
Molecular Weight:
349.42

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1