![Avicularin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/561/477/caa4855c-f930-46b3-ad74-bcb41f0e4dcf/640/caa4855c-f930-46b3-ad74-bcb41f0e4dcf.png)
Avicularin
![Avicularin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/561/477/caa4855c-f930-46b3-ad74-bcb41f0e4dcf/640/caa4855c-f930-46b3-ad74-bcb41f0e4dcf.png)
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-O-α-L-arabinofuranoside, Avicularoside, Quercetin 3-O-α-L-arabinofuranoside
Empirical Formula (Hill Notation):
C20H18O11
CAS Number:
Molecular Weight:
434.35
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing