
Ascorbyl Glucoside

Synonym(s):
Ascorbyl Glucoside
Empirical Formula (Hill Notation):
C12H18O11
CAS Number:
Molecular Weight:
338.26
Products
Product Category
Brand
Formula Weight
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing