Skip to Content
Merck
Arctiin phyproof® Reference Substance

Arctiin

Arctiin phyproof® Reference Substance
Synonym(s):
Arctigenin 4-glucoside
Empirical Formula (Hill Notation):
C27H34O11
CAS Number:
Molecular Weight:
534.55

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing