
Anti-YAP antibody produced in rabbit

Synonym(s):
65 kDa Yes-associated protein, YAP1, YAP65
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing