
Anti-PITRM1 antibody produced in rabbit

Synonym(s):
Anti-Metalloprotease 1 antibody produced in rabbit, Anti-Pitrilysin metalloproteinase 1 antibody produced in rabbit, Anti-Presequence protease, mitochondrial precursor antibody produced in rabbit, Anti-hMP1 antibody produced in rabbit, Anti-hPreP antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing