![Anti-IGBP1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/340/941/32bd2cd6-ce80-478d-979a-3274022906df/640/32bd2cd6-ce80-478d-979a-3274022906df.jpg)
Anti-IGBP1 antibody produced in rabbit
![Anti-IGBP1 antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/340/941/32bd2cd6-ce80-478d-979a-3274022906df/640/32bd2cd6-ce80-478d-979a-3274022906df.jpg)
Synonym(s):
Anti-α 4 Protein antibody produced in rabbit, Anti-α4 Protein antibody produced in rabbit, Anti-B cell signal transduction molecule α 4 antibody produced in rabbit, Anti-B cell signal transduction molecule α4 antibody produced in rabbit, Anti-CD79a-binding protein 1 antibody produced in rabbit, Anti-Immunoglobulin-binding protein 1 antibody produced in rabbit, Anti-Renal carcinoma antigen NY-REN-16 antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing