
Anti-Caveolin-1 antibody produced in rabbit

Synonym(s):
Anti-Caveolin-1, Anti-VIP21, CAV, CAV1, Caveolin-1
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing