
Antazoline phosphate

Synonym(s):
Antazoline phosphate salt, 2-(N-Benzylanilinomethyl)-2-imidazoline
Linear Formula:
C17H19N3 · H3PO4
CAS Number:
Molecular Weight:
363.35
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2