![Anisyl butyrate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/407/307/517a44f1-5fc7-417b-9c18-007da2746017/640/517a44f1-5fc7-417b-9c18-007da2746017.png)
Anisyl butyrate
![Anisyl butyrate United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/407/307/517a44f1-5fc7-417b-9c18-007da2746017/640/517a44f1-5fc7-417b-9c18-007da2746017.png)
Synonym(s):
p-Methoxybenzyl butyrate
Empirical Formula (Hill Notation):
C12H16O3
CAS Number:
Molecular Weight:
208.25
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing