![Amitraz Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/901/503/df048132-eac4-4675-a050-effeb27ecdb1/640/df048132-eac4-4675-a050-effeb27ecdb1.jpg)
Amitraz Related Compound C
![Amitraz Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/901/503/df048132-eac4-4675-a050-effeb27ecdb1/640/df048132-eac4-4675-a050-effeb27ecdb1.jpg)
Synonym(s):
N,N′-Bis(2,4-xylyl)formamidine, N,N′-Bis(2,4-dimethylphenyl)methanimidamide
Empirical Formula (Hill Notation):
C17H20N2
CAS Number:
Molecular Weight:
252.35
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing