![Amitraz Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/255/131/ae14481b-745e-40b4-8d65-ff6a4afbd3d9/640/ae14481b-745e-40b4-8d65-ff6a4afbd3d9.png)
AMITRAZ RELATED COMPOUND B
![Amitraz Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/255/131/ae14481b-745e-40b4-8d65-ff6a4afbd3d9/640/ae14481b-745e-40b4-8d65-ff6a4afbd3d9.png)
Synonym(s):
Amitraz Metabolite BTS 27271, N-(2,4-Dimethylphenyl)-N′-methylformamidine, N-Methyl-N′-2,4-xylylformamidine
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2