Skip to Content
Merck
Aloe-emodin analytical standard

Aloe-emodin

Aloe-emodin analytical standard
Synonym(s):
1,8-Dihydroxy 3-hydroxymethylanthraquinone, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone, 3-Hydroxymethylchrysazine, Rhabarberone
Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Tissue
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing