
Allopurinol Related Compound F

Synonym(s):
Ethyl (E/Z)-3-(2-carbethoxy-2-cyanoethenyl)amino-1H-pyrazole-4-carboxylate
Empirical Formula (Hill Notation):
C12H14N4O4
CAS Number:
Molecular Weight:
278.26
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2