Adapalene
Synonym(s):
6-(4-Methoxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-2-naphthalenecarboxylic acid, 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic acid, CD-271, Differin, Adapalene
Empirical Formula (Hill Notation):
C28H28O3
CAS Number:
Molecular Weight:
412.52
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing